missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-8 / CXCL8 Protein

A DNA sequence encoding the Human IL-8 / CXCL8, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10315-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923478

Cantidad Precio
1 105.00€ / 5 microgramos
10 98.00€ / 5 microgramos
Fecha estimada de envío
10-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_000575.1
IL-8 / CXCL8 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SAKELRCQCI KTYSKPFHPK FIKELRVIES GPHCANTEII VKLSDGRELC LDPKENWVQR VVEKFLKRAE NS
3576
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL8
The ED50 as determined by a chemotaxis bioassay using human CXCR2 transfected mouse BaF3 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos