missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-8 / CXCL8 Protein
Recombinant Protein
Marca: enQuireBio™ QP2737-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000575.1 | |
IL-8 / CXCL8 Protein | |
Research Use Only | |
CXCL8 | |
Recombinant Protein | |
E. coli | |
AVLPRSAKEL . RCQCIKTYS K. PFHPKFIK EL. RVIESGP HCA. NTEIIV KLSD. GRELC LDPKE. NWVQ RVVEKF. LKR AENS | |
Greater than 95% as determined by SDS-PAGE. |
3576 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human IL-8 / CXCL8 induces Human neutrophil chemotaxis (significanty different from control at Human IL-8 / CXCL8 concentrations greater than 10 ng/ml) Activity was further confirmed by measuring Human neutrophil degranulation (significanty different from control at Human IL-8 / CXCL8 concentrations greater than 1 ng/ml) . | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |