missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-3 / Interleukin-3 Protein
A DNA sequence encoding the Human IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10310-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000579.2 | |
IL-3 / Interleukin-3 Protein | |
Research Use Only | |
IL3 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3562 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APMTQTTSLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF | |
Greater than 96% as determined by SDS-PAGE and HPLC. |