missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-15 Protein

A DNA sequence encoding the Homo sapiens (Human) IL-15, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8555-ye-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055251

  • 530.00€ / 100 microgramos


Fecha estimada de envío
16-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000576.1
IL-15 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3600
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL15
Recombinant Protein
Yeast
NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SLESGDASIH DTVENLIILA NNSLSSNGNV TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS
Videos
Documentos