missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-15 Protein
A DNA sequence encoding the Human IL-15, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10302-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000576.1 | |
IL-15 Protein | |
Research Use Only | |
IL15 | |
The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3600 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SLESGDASIH DTVENLIILA NNSLSSNGNV TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |