missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IgG receptor FcRn large subunit p51 Protein

A DNA sequence encoding the Homo sapiens (Human) IgG receptor FcRn large subunit p51, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6030-ma-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909489

  • 1014.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_001129491.1
IgG receptor FcRn large subunit p51 Protein
50 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
2217
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FCGRT
Recombinant Protein
Mammal
AESHLSLLYH LTAVSSPAPG TPAFWVSGWL GPQQYLSYNS LRGEAEPCGA WVWENQVSWY WEKETTDLRI KEKLFLEAFK ALGGKGPYTL QGLLGCELGP DNTSVPTAKF ALNGEEFMNF DLKQGTWGGD WPEALAISQR WQQQDKAANK ELTFLLFSCP HRLREHLERG RGNLEWKEPP SMRLKARPSS PGFSVLTCSA FSFYPPELQL RFLRNGLAAG TGQGDFGPNS DGSFHASSSL TVKSGDEHHY CCIVQHAGLA QPLRVELESP AKSS
Videos