missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGFBP3 Protein

A DNA sequence encoding the Human IGFBP3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10386-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15943698

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Especificaciones

Especificaciones

NP_000589.2
IGFBP3 Protein
Research Use Only
IGFBP3
The ED50 as determined by inhibiting IGF-II induced proliferation of serum free human MCF-7 cells is less than 200 ng/ml, corresponding to a specific activity of > 5.0 M 103 IU/mg in the presence of 15 ng/ml of rHuIGF-II.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
3486
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GASSGGLGPV VRCEPCDARA LAQCAPPPAV CAELVREPGC GCCLTCALSE GQPCGIYTER CGSGLRCQPS PDEARPLQAL LDGRGLCVNA SAVSRLRAYL LPAPPAPGNA SESEEDRSAG EVESPSVSST HRVSDPKFHP LHSKIIIIKK GHAKDSQRYK VDYESQSTDT QNFSSESKRE TEYGPCRREM EDTLNHLKFL NVLSPRGVHI PNCDKKGFYK KKQCRPSKGR KRGFCWCVDK YGQPLPGYTT KGKEDVHCYS MQSK
Greater than 98% as determined by SDS-PAGE and HPLC.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales