missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN a 1b Protein
A cDNA sequence encoding the IFN a 1b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10673-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
3439; 3447 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity as determined in a viral resistance assay using bovine kidney MDBK cells was found to be 1 x 108 IU/ mg. | |
Human | |
Untagged | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IFN a 1b Protein | |
Research Use Only | |
IFNA1; IFNA13 | |
Recombinant Protein | |
E. coli | |
Lyophilized from a 0.2microm filtered solution (50ug/ml) in PBS, pH 7.4, containing 4% mannitol and 1% HSA. |