missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IDS Protein

A cDNA sequence encoding the IDS was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12369-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920199

  • 5941.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 08-04-2019
Añadir a la cesta

Especificación

Especificación

IDS Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
IDS protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
SETQANSTTD ALNVLLIIVD DLRPSLGCYG DKLVRSPNID QLASHSLLFQ NAFAQQAVCA PSRVSFLTGR RPDTTRLYDF NSYWRVHAGN FSTIPQYFKE NGYVTMSVGK VFHPGISSNH TDDSPYSWSF PPYHPSSEKY ENTKTCRGPD GELHANLLCP VDVLDVPEGT LPDKQSTEQA IQLLEKMKTS ASPFFLAVGY HKPHIPFRYP KEFQKLYPLE NITLAPDPEV PDGLPPVAYN PWMDIRQRED VQALNISVPY GPIPVDFQRK IRQSYFASVS YLDTQVGRLL SALDDLQLAN STIIAFTSDH GWALGEHGEW AKYSNFDVAT HVPLIFYVPG RTASLPEAGE KLFPYLDPFD SASQLMEPGR QSMDLVELVS LFPTLAGLAG LQVPPRCPVP SFHVELCREG KNLLKHFRFR DLEEDPYLPG NPRELIAYSQ YPRPSDIPQW NSDKPSLKDI KIMGYSIRTI DYRYTVWVGF NPDEFLANFS DIHAGELYFV DSDPLQDHNM YNDSQGGDLF QLLMPLEHHH HHH
Videos