missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human I-309 / CCL1 / TCA-3 Protein

A DNA sequence encoding the Human I-309 / CCL1 / TCA-3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10223-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923138

  • 1153.00€ / 250 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002972.1
I-309 / CCL1 / TCA-3 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK
6346
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL1
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 10-100 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos