Learn More
enQuireBio™ Recombinant Human I-309 / CCL1 / TCA-3 Protein
A DNA sequence encoding the Human I-309 / CCL1 / TCA-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10223-ec-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002972.1 | |
I-309 / CCL1 / TCA-3 Protein | |
Research Use Only | |
CCL1 | |
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6346 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK | |
Greater than 97% as determined by SDS-PAGE and HPLC. |