missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HYKK Protein
A cDNA sequence encoding the HYKK was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12362-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HYKK Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSGNYQ QSEALSKPTF SEEQASALVE SVFGLKVSKV RPLPSYDDQN FHVYVSKTKD GPTEYVLKIS NTKASKNPDL IEVQNHIIMF LKAAGFPTAS VCHTKGDNTA SLVSVDSGSE IKSYLVRLLT YLPGRPIAEL PVSPQLLYEI GKLAAKLDKT LQRFHHPKLS SLHRENFIWN LKNVPLLEKY LYALGQNRNR EIVEHVIHLF KEEVMTKLSH FRECINHGDL NDHNILIESS KSASGNAEYQ VSGILDFGDM SYGYYVFEVA ITIMYMMIES KSPIQVGGHV LAGFESITPL TAVEKGALFL LVCSRFCQSL VMAAYSCQLY PENKDYLMVT AKTGWKHLQQ MFDMGQKAVE EIWFETAKSY ESGISM | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HYKK solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4), 40% glycerol, 1mM DTT and 0.1mM PMSF. |