missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HVEM / TNFRSF14 Protein

A DNA sequence encoding the Human HVEM / TNFRSF14, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10371-ye-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923658

  • 357.00€ / 250 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_003811.2
HVEM / TNFRSF14 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
Yeast
LPSCKEDEYP VGSECCPKCS PGYRVKEACG ELTGTVCEPC PPGTYIAHLN GLSKCLQCQM CDPAMGLRAS RNCSRTENAV CGCSPGHFCI VQDGDHCAAC RAYATSSPGQ RVQKGGTESQ DTLCQNCPPG TFSPNGTLEE CQHQTKCSWL VTKAGAGTSS SHWVEPKSSD KTHTCPPCPA PEFEGAPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPTP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK
8764
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
TNFRSF14
The biologically active as determined by its ability to inhibit TNF-beta -mediated cytotoxicity using murine L929 cells.
Human
FC
0.2 um filtered PBS, pH7.4, with 3 % Trehalose, lyophilized
Videos