missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HTF Protein

A cDNA sequence encoding the HTF was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12352-5gram

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15920149

  • 1833.00€ / 5 g


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

7018
Greater than 98.0% as determined by coomassie blue stained SDS-PAGE and Cellulose Acetate electrophoresis.
Research Use Only
TF
Human
Untagged
The protein (10 mg/ml) was lyophilized from 20mM NH4HC03 solution. May contain traces of buffer salts.
HTF Protein
5 g
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Native Protein
MRLAVGALLV CAVLGLCLAV PDKTVRWCAV SEHEATKCQS FRDHMKSVIP SDGPSVACVK KASYLDCIRA IAANEADAVT LDAGLVYDAY LAPNNLKPVV AEFYGSKEDP QTFYYAVAVV KKDSGFQMNQ LRGKKSCHTG LGRSAGWNIP IGLLYCDLPE PRKPLEKAVA NFFSGSCAPC ADGTDFPQLC QLCPGCGCST LNQYFGYSGA FKCLKDGAGD VAFVKHSTIF ENLANKADRD QYELLCLDNT RKPVDEYKDC HLAQVPSHTV VARSMGGKED LIWELLNQAQ EHFGKDKSKE FQLFSSPHGK DLLFKDSAHG FLKVPPRMDA KMYLGYEYVT AIRNLREGTC PEAPTDECKP VKWCALSHHE RLKCDEWSVN SVGKIECVSA ETTEDCIAKI MNGEADAMSL DGGFVYIAGK CGLVPVLAEN YNKSDNCEDT PEAGYFAIAV VKKSASDLTW DNLKGKKSCH TAVGRTAGWN IPMGLLYNKI NHCRFDEFFS EGCAPGSKKD SSLCKLCMGS GLNLCEPNNK EGYYGYTGAF RCLVEKGDVA FVKHQTVPQN TGGKNPDPWA KNLNEKDYEL LCLDGTRKPV EEYANCHLAR APNHAVVTRK DKEACVHKIL RQQQHLFGSN VTDCSGNFCL FRSETKDLLF RDDTVCLAKL HDRNTYEKYL GEEYVKAVGN LRKCSTSSLL EACTFRRP
Videos