missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSD11B1 Protein

A DNA sequence encoding the Homo sapiens (Human) HSD11B1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6183-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051390

  • 406.00€ / 100 microgramos


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001193670.1
HSD11B1 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3290
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HSD11B1
Recombinant Protein
E. coli
EEFRPEMLQG KKVIVTGASK GIGREMAYHL AKMGAHVVVT ARSKETLQKV VSHCLELGAA SAHYIAGTME DMTFAEQFVA QAGKLMGGLD MLILNHITNT SLNLFHDDIH HVRKSMEVNF LSYVVLTVAA LPMLKQSNGS IVVVSSLAGK VAYPMVAAYS ASKFALDGFF SSIRKEYSVS RVNVSITLCV LGLIDTETAM KAVSGIVHMQ AAPKEECALE IIKGGALRQE EVYYDSSLWT TLLIRNPCRK ILEFLYSTSY NMDRFINK
Videos
Documentos