missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HRAS Protein
A cDNA sequence encoding the HRAS was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12311-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HRAS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTEYKLVVVG AGGVGKSALT IQLIQNHFVD EYDPTIEDSY RKQVVIDGET CLLDILDTAG QEEYSAMRDQ YMRTGEGFLC VFAINNTKSF EDIHQYREQI KRVKDSDDVP MVLVGNKCDL AARTVESRQA QDLARSYGIP YIETSAKTRQ GVEDAFYTLV REIRQHKLRK LNPPDESGPG CMSCKCLEHH HHHH | |
Greater than 90% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
0.5 mg/ml solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 20% glycerol. |