missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HNRNPAB Protein

A cDNA sequence encoding the HNRNPAB was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12290-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909958

  • 103.00€ / 2 microgramos


Fecha estimada de envío
12-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

HNRNPAB Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HNRNPAB protein solution (0.25 mg/ml) containing 30% glycerol and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSEAGEE QPMETTGATE NGHEAVPEGE SPAGAGTGAA AGAGGATAAP PSGNQNGAEG DQINASKNEE DAGKMFVGGL SWDTSKKDLK DYFTKFGEVV DCTIKMDPNT GRSRGFGFIL FKDAASVEKV LDQKEHRLDG RVIDPKKAMA MKKDPVKKIF VGGLNPEATE EKIREYFGEF GEIEAIELPM DPKLNKRRGF VFITFKEEEP VKKVLEKKFH TVSGSKCEIK VAQPKEVYQQ QQYGSGGRGN RNRGNRGSGG GGGGGGQGST NYGKSQRRGG HQNNYKPY
Videos