missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGCS1 Protein
A cDNA sequence encoding the HMGCS1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12284-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
3157 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPGSLPL NAEACWPKDV GIVALEIYFP SQYVDQAELE KYDGVDAGKY TIGLGQAKMG FCTDREDINS LCMTVVQNLM ERNNLSYDCI GRLEVGTETI IDKSKSVKTN LMQLFEESGN TDIEGIDTTN ACYGGTAAVF NAVNWIESSS WDGRYALVVA GDIAVYATGN ARPTGGVGAV ALLIGPNAPL IFERGLRGTH MQHAYDFYKP DMLSEYPIVD GKLSIQCYLS ALDRCYSVYC KKIHAQWQKE GNDKDFTLND FGFMIFHSPY CKLVQKSLAR MLLNDFLNDQ NRDKNSIYSG LEAFGDVKLE DTYFDRDVEK AFMKASSELF SQKTKASLLV SNQNGNMYTS SVYGSLASVL AQYSPQQLAG KRIGVFSYGS GLAATLYSLK VTQDATPGSA LDKITASLCD LKSRLDSRTG VAPDVFAENM KLREDTHHLV NYIPQGSIDS LFEGTWYLVR VDEKHRRTYA RRPTPNDDTL DEGVGLVHSN IATEHIPSPA KKVPRLPATA AEPEAAVISN GEH | |
Greater than 80.0% as determined by SDS-PAGE. |
HMGCS1 Protein | |
Research Use Only | |
HMGCS1 | |
Human | |
His | |
HMGCS1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |