missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HMGA2 Protein

A cDNA sequence encoding the HMGA2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12281-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919928

  • 1372.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

8091
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
HMGA2
Human
His
HMGA2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 250mM Imidazole.
HMGA2 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSARGEGAGQ PSTSAQGQPA APAPQKRGRG RPRKQQQEPT GEPSPKRPRG RPKGSKNKSP SKAAQKKAEA TGEKRPRGRP RKWPQQVVQK KPAQEETEET SSQESAEEDL EHHHHHH
Videos