missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGA1 Protein
A cDNA sequence encoding the HMGA1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12280-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HMGA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSESSSKSSQ PLASKQEKDG TEKRGRGRPR KQPPVSPGTA LVGSQKEPSE VPTPKRPRGR PKGSKNKGAA KTRKTTTTPG RKPRGRPKKL EKEEEEGISQ ESSEEEQLEH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HMGA1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50% glycerol and 0.2M NaCl. |