missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2B type 1-B Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6159-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_066406.1 | |
Histone H2B type 1-B Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3018 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
HIST1H2BB | |
Recombinant Protein | |
E. coli | |
PEPSKSAPAP KKGSKKAITK AQKKDGKKRK RSRKESYSIY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSSK |