missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human High affinity copper uptake protein 1

A DNA sequence encoding the Homo sapiens (Human) High affinity copper uptake protein 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9957-iv-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16059091

  • 1725.00€ / 100 microgramos


Fecha estimada de envío
19-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001850.1
High affinity copper uptake protein 1
100 μg
Not determined.
Full Length Transmembrane Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
1317
Greater than 85% as determined by SDS-PAGE.
Research Use Only
SLC31A1
Recombinant Protein
Invitro-E. coli
MDHSHHMGMS YMDSNSTMQP SHHHPTTSAS HSHGGGDSSM MMMPMTFYFG FKNVELLFSG LVINTAGEMA GAFVAVFLLA MFYEGLKIAR ESLLRKSQVS IRYNSMPVPG PNGTILMETH KTVGQQMLSF PHLLQTVLHI IQVVISYFLM LIFMTYNGYL CIAVAAGAGT GYFLFSWKKA VVVDITEHCH
Videos
Documentos