missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HES2 Protein

A cDNA sequence encoding the HES2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12219-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909738

  • 5941.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

HES2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HES2 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGLPRRA GDAAELRKSL KPLLEKRRRA RINQSLSQLK GLILPLLGRE NSNCSKLEKA DVLEMTVRFL QELPASSWPT AAPLPCDSYR EGYSACVARL ARVLPACRVL EPAVSARLLE HLWRRAASAT LDGGRAGDSS GPSAPAPAPA SAPEPASAPV PSPPSPPCGP GLWRPW
Videos