missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hermansky-Pudlak syndrome 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Hermansky-Pudlak syndrome 1 protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7996-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000186.2 | |
Hermansky-Pudlak syndrome 1 Protein | |
Research Use Only | |
HPS1 | |
Recombinant Protein | |
E. coli | |
MKCVLVATEG AEVLFYWTDQ EFEESLRLKF GQSENEEEEL PALEDQLSTL LAPVIISSMT MLEKLSDTYT CFSTENGNFL YVLHLFGECL FIAINGDHTE SEGDLRRKLY VLKYLFEVHF GLVTVDGHLI RKELRPPDLA QRVQLWEHFQ SLLWTYSRLR EQEQCFAVEA LERLIHPQLC ELCIEALERH VIQAVNTSPE RGGEEALHAF LLVHSKLLAF YSSHSASSLR PADLLALILL VQDLYPSEST AEDDIQPSPR RARSSQNIPV QQAWSPHSTG PTGGSSAETE TDSFSLPEEY FTPAPSPGDQ SSGEDRRKAG GNNS | |
Greater than 80% as determined by SDS-PAGE. |
3257 | |
100 μg | |
Not determined. | |
Full Length of Isoform III | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |