missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HER2 / ErbB2 Protein

A DNA sequence encoding the Homo sapiens (Human) HER2 / ErbB2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5993-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929209

  • 675.00€ / 200 microgramos


Fecha estimada de envío
07-08-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001005862.1
HER2 / ErbB2 Protein
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
2064
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ERBB2
Recombinant Protein
E. coli
TQVCTGTDMK LRLPASPETH LDMLRHLYQG CQVVQGNLEL TYLPTNASLS FLQDIQEVQG YVLIAHNQVR QVPLQRLRIV RGTQLFEDNY ALAVLDNGDP LNNTTPVTGA SPGGLRELQL RSLTEILKGG VLIQRNPQLC YQDTILWKDI FHKNNQLALT LIDTNRSRAC HPCSPMCKGS RCWGESSEDC QSLTRTVCAG GCARCKGPLP TDCCHEQCAA GCTGPKHSDC LACLHFNHSG ICELHCPALV TYNTDTFESM PNPEGRYTFG ASCVTACPYN YLSTDVGSCT LVCPLHNQEV TAEDGTQRCE KCSKPCARVC YGLGMEHLRE VRAVTSANIQ EFAGCKKIFG SLAFLPESFD GDPASNTAPL QPEQLQVFET LEEITGYLYI SAWPDSLPDL SVFQNLQVIR GRILHNGAYS LTLQGLGISW LGLRSLRELG SGLALIHHNT HLCFVHTVPW DQLFRNPHQA LLHTANRPED ECVGEGLACH QLCARGHCWG PGPTQCVNCS QFLRGQECVE ECRVLQGLPR EYVNARHCLP CHPECQPQNG SVTCFGPEAD QCVACAHYKD PPFCVARCPS GVKPDLSYMP IWKFPDEEGA CQPCPINCTH SCVDLDDKGC PAEQRASPLT
Videos
Documentos