missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HDL Protein

A cDNA sequence encoding the HDL was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12211-100mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929708

  • 710.00€ / 100 miligramos


Fecha estimada de envío
18-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

HDL Protein
100 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Cholesterol level >200 mg/l. <1% LDL.
Research Use Only
Recombinant Protein
Native Protein
Videos
Documentos