missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDAC8 / HDACL1 Protein
A DNA sequence encoding the Homo sapiens (Human) HDAC8 / HDACL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6155-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001159890.1 | |
HDAC8 / HDACL1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
55869 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
HDAC8 | |
Recombinant Protein | |
E. coli | |
MEEPEEPADS GQSLVPVYIY SPEYVSMCDS LAKIPKRASM VHSLIEAYAL HKQMRIVKPK VASMEEMATF HTDAYLQHLQ KVSQEGDDDH PDSIEYGLGY DCPATEGIFD YAAAIGGATI TAAQCLIDGM CKVAINWSGG WHHAKKDEAS GFCYLNDAVL GILRLRRKFE RILYVDLDLH HGDGVEDAFS FTSKVMTVSL HKFSPGFFPG TGDVSDVGLG KGRYYSVNVP IQDGIQDEKY YQICESVLKE VYQAFNPKAV VLQLGADTIA GDPMCSFNMT PVGIGKCLKY ILQWQLATLI LGGGGYNLAN TARCWTYLTG VILGKTLSSE IPDHEFFTAY GPDYVLEITP SCRPDRNEPH RIQQILNYIK GNLKHVV |