missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HBEGF / DTR Protein

A DNA sequence encoding the Human HBEGF / DTR Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10400-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923738

  • 333.00€ / 100 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001936.1
HBEGF / DTR Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
DLQEADLDLL RVTLSSKPQA LATPNKEEHG KRKKKGKGLG KKRDPCLRKY KDFCIHGECK YVKELRAPSC ICHPGYHGER CHGLSL
1839
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
HBEGF
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized
Videos