missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HADH Protein
A cDNA sequence encoding the HADH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12099-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
3033 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSSSTASAS AKKIIVKHVT VIGGGLMGAG IAQVAAATGH TVVLVDQTED ILAKSKKGIE ESLRKVAKKK FAENPKAGDE FVEKTLSTIA TSTDAASVVH STDLVVEAIV ENLKVKNELF KRLDKFAAEH TIFASNTSSL QITSIANATT RQDRFAGLHF FNPVPVMKLV EVIKTPMTSQ KTFESLVDFS KALGKHPVSC KDTPGFIVNR LLVPYLMEAI RLYERGDASK EDIDTAMKLG AGYPMGPFEL LDYVGLDTTK FIVDGWHEMD AENPLHQPSP SLNKLVAENK FGKKTGEGFY KYK | |
Greater than 95% as determined by SDS-PAGE. |
HADH Protein | |
Research Use Only | |
HADH | |
Human | |
His | |
The HADH solution contains 20mM Tris-HCl pH 8.0, 0.1M NaCl and 20% glycerol. |