missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GULP1 Protein

A cDNA sequence encoding the GULP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12069-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909268

  • 160.00€ / 20 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

GULP1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GULP1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol and 2mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMNRAFSR KKDKTWMHTP EALSKHFIPY NAKFLGSTEV EQPKGTEVVR DAVRKLKFAR HIKKSEGQKI PKVELQISIY GVKILEPKTK EVQHNCQLHR ISFCADDKTD KRIFTFICKD SESNKHLCYV FDSEKCAEEI TLTIGQAFDL AYRKFLESGG KDVETRKQIA GLQKRIQDLE TENMELKNKV QDLENQLRIT QVSAPPAGSM TPKSPSTDIF DMIPFSPISH QSSMPTRNGT QPPPVPSRST EIKRDLFGAE PFDPFNCGAA DFPPDIQSKL DEMQEGFKMG LTLEGTVFCL DPLDSRC
Videos