missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GTF2E2 Protein

A DNA sequence encoding the Homo sapiens (Human) GTF2E2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6131-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051030

  • 406.00€ / 100 microgramos


Fecha estimada de envío
27-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002086.1
GTF2E2 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2961
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GTF2E2
Recombinant Protein
E. coli
MDPSLLRERE LFKKRALSTP VVEKRSASSE SSSSSSKKKK TKVEHGGSSG SKQNSDHSNG SFNLKALSGS SGYKFGVLAK IVNYMKTRHQ RGDTHPLTLD EILDETQHLD IGLKQKQWLM TEALVNNPKI EVIDGKYAFK PKYNVRDKKA LLRLLDQHDQ RGLGGILLED IEEALPNSQK AVKALGDQIL FVNRPDKKKI LFFNDKSCQF SVDEEFQKLW RSVTVDSMDE EKIEEYLKRQ GISSMQESGP KKVAPIQRRK KPASQKKRRF KTHNEHLAGV LKDYSDITSS K
Videos
Documentos