missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSTM1 Protein

A cDNA sequence encoding the GSTM1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12057-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919228

  • 160.00€ / 10 microgramos


Fecha estimada de envío
18-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

GSTM1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GSTM1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH RSMPMILGYW DIRGLAHAIR LLLEYTDSSY EEKKYTMGDA PDYDRSQWLN EKFKLGLDFP NLPYLIDGAH KITQSNAILC YIARKHNLCG ETEEEKIRVD ILENQTMDNH MQLGMICYNP EFEKLKPKYL EELPEKLKLY SEFLGKRPWF AGNKGLEKIS AYMKSSRFLP RPVFSKMAVW GNK
Videos