missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GRP94 / HSP90B1 Protein
A DNA sequence encoding the Homo sapiens (Human) GRP94 / HSP90B1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8707-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003290.1 | |
GRP94 / HSP90B1 Protein | |
Research Use Only | |
HSP90B1 | |
Recombinant Protein | |
E. coli | |
VDGTVEEDLG KSREGSRTDD EVVQREEEAI QLDGLNASQI RELREKSEKF AFQAEVNRMM KLIINSLYKN KEIFLRELIS NASDALDKIR LISLTDENAL SGNEELTVKI KCDKEKNLLH VTDTGVGMTR EELVKNLGTI AKSGTSEFLN KMTEAQEDGQ STSELIGQFG VGFYSAFLVA DKVIVTSKHN NDTQHIWESD SNEFSVIADP RGNTLGRGTT ITLVLKEEAS DYLELDTIKN LVKKYSQFIN FPIYVWSSKT ETVEEPMEEE EAAKEEKEES DDEAAVEEEE EEKKPKTKKV EKTVWDWELM NDIKPIWQRP SKEVEEDEYK AFYKSFSKES DDPMAYIHFT AEGEVTFKSI LFVPTSAPRG LFDEYGSKKS DYIKLYVRRV FITDDFHDMM PKYLNFVKGV VDSDDLPLNV SRETLQQHKL LKVIRKKLVR KTLDMIKKIA DDKYNDTFWK EFGTNIKLGV IEDHSNRTRL AKLLRFQSSH HPTDITSLDQ YVERMKEKQD KIYFMAGSSR KEAESSPFVE RLLKKGYEVI YLTEPVDEYC IQALPEFDGK RFQNVAKEGV KFDESEKTKE SREAVEKEFE PLLNWMKDKA LKDKIEKAVV SQRLTESPCA LVASQYGWSG NMERIMKAQA YQTGKDISTN YYASQKKTFE INPRHPLIRD MLRRIKEDED DKTVLDLAVV LFETATLRSG YLLPDTKAYG DRIERMLRLS LNIDPDAKVE EEPEEEPEET AEDTTEDTEQ DEDEEMDVGT DEEEETAKES TAE | |
Greater than 90% as determined by SDS-PAGE. |
7184 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |