missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GOLM1 / GP73 Protein
A DNA sequence encoding the Homo sapiens (Human) GOLM1 / GP73, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8697-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_057632.2 | |
GOLM1 / GP73 Protein | |
Research Use Only | |
GOLM1 | |
Recombinant Protein | |
E. coli | |
SSRSVDLQTR IMELEGRVRR AAAERGAVEL KKNEFQGELE KQREQLDKIQ SSHNFQLESV NKLYQDEKAV LVNNITTGER LIRVLQDQLK TLQRNYGRLQ QDVLQFQKNQ TNLERKFSYD LSQCINQMKE VKEQCEERIE EVTKKGNEAV ASRDLSENND QRQQLQALSE PQPRLQAAGL PHTEVPQGKG NVLGNSKSQT PAPSSEVVLD SKRQVEKEET NEIQVVNEEP QRDRLPQEPG REQVVEDRPV GGRGFGGAGE LGQTPQVQAA LSVSQENPEM EGPERDQLVI PDGQEEEQEA AGEGRNQQKL RGEDDYNMDE NEAESETDKQ AALAGNDRNI DVFNVEDQKR DTINLLDQRE KRNHTL | |
Greater than 90% as determined by SDS-PAGE. |
51280 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |