missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNA15 Protein

A DNA sequence encoding the Homo sapiens (Human) GNA15, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6100-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050800

  • 1715.00€ / 500 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002059.3
GNA15 Protein
500 μg
Not determined.
Full Length
Human
His-Trx
Tris-based buffer, 50% glycerol
2769
Greater than 80% as determined by SDS-PAGE.
Research Use Only
GNA15
Recombinant Protein
E. coli
MARSLTWRCC PWCLTEDEKA AARVDQEINR ILLEQKKQDR GELKLLLLGP GESGKSTFIK QMRIIHGAGY SEEERKGFRP LVYQNIFVSM RAMIEAMERL QIPFSRPESK HHASLVMSQD PYKVTTFEKR YAAAMQWLWR DAGIRAYYER RREFHLLDSA VYYLSHLERI TEEGYVPTAQ DVLRSRMPTT GINEYCFSVQ KTNLRIVDVG GQKSERKKWI HCFENVIALI YLASLSEYDQ CLEENNQENR MKESLALFGT ILELPWFKST SVILFLNKTD ILEEKIPTSH LATYFPSFQG PKQDAEAAKR FILDMYTRMY TGCVDGPEGS KKGARSRRLF SHYTCATDTQ NIRKVFKDVR DSVLARYLDE INLL
Videos
Documentos