missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GM-CSF / CSF2 Protein

A DNA sequence encoding the Homo sapiens (Human) GM-CSF / CSF2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5876-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928389

  • 345.00€ / 50 microgramos


Fecha estimada de envío
17-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000749.2
GM-CSF / CSF2 Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
1437
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CSF2
Recombinant Protein
Yeast
APARSPSPST QPWEHVNAIQ EARRLLNLSR DTAAEMNETV EVISEMFDLQ EPTCLQTRLE LYKQGLRGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFESFKENL KDFLLVIPFD CWEPVQE
Videos
Documentos