missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glycerol kinase 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Glycerol kinase 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6085-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919859

  • 183.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

NP_149991.2
Glycerol kinase 2 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2712
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GK2
Recombinant Protein
E. coli
AAPKTAAVGP LVGAVVQGTN STRFLVFNSK TAELLSHHKV ELTQEFPKEG WVEQDPKEIL QSVYECIART CEKLDELNID ISNIKAVGVS NQRETTVIWD KLTGEPLYNA VVWLDLRTQT TVEDLSKKIP GNSNFVKSKT GLPLSTYFSA VKLRWMLDNV RNVQKAVEEG RALFGTIDSW LIWSLTGGVN GGVHCTDVTN ASRTMLFNIH SLEWDKELCD FFEIPMDLLP NVFSSSEIYG LIKTGALEGV PISGCLGDQC AALVGQMCFQ EGQAKNTYGT GCFLLCNTGR KCVFSEHGLL TTVAYKLGRE KPAYYALEGS VAIAGAVIRW LRDNLGIIET SGDIERLAKE VGTSYGCYFV PAFSGLYAPY WEPSARGILC GLTQFTNKCH IAFAALEAVC FQTREILEAM NRDCGIPLRH LQVDGGMTNN KVLMQLQADI LHIPVIKPFM PETTALGAAM AAGAAEGVSV WSLEPQALSV LRMERFEPQI QATESEIRYA TWKKAVMKSM GWVTSQSPEG GDPSIFSSLP LGFFIVSSMV MLIGARYISG VP
Videos