missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutaredoxin-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin-3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6094-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050770

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
27-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001186797.1
Glutaredoxin-3 Protein
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
10539
Greater than 80% as determined by SDS-PAGE.
Research Use Only
GLRX3
Recombinant Protein
E. coli
AAGAAEAAVA AVEEVGSAGQ FEELLRLKAK SLLVVHFWAP WAPQCAQMNE VMAELAKELP QVSFVKLEAE GVPEVSEKYE ISSVPTFLFF KNSQKIDRLD GAHAPELTKK VQRHASSGSF LPSANEHLKE DLNLRLKKLT HAAPCMLFMK GTPQEPRCGF SKQMVEILHK HNIQFSSFDI FSDEEVRQGL KAYSSWPTYP QLYVSGELIG GLDIIKELEA SEELDTICPK APKLEERLKV LTNKASVMLF MKGNKQEAKC GFSKQILEIL NSTGVEYETF DILEDEEVRQ GLKAYSNWPT YPQLYVKGEL VGGLDIVKEL KENGELLPIL RGEN
Videos
Documentos