missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GID8 Protein
A cDNA sequence encoding the GID8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11970-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GID8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSYAEKP DEITKDEWME KLNNLHVQRA DMNRLIMNYL VTEGFKEAAE KFRMESGIEP SVDLETLDER IKIREMILKG QIQEAIALIN SLHPELLDTN RYLYFHLQQQ HLIELIRQRE TEAALEFAQT QLAEQGEESR ECLTEMERTL ALLAFDSPEE SPFGDLLHTM QRQKVWSEVN QAVLDYENRE STPKLAKLLK LLLWAQNELD QKKVKYPKMT DLSKGVIEEP K | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GID8 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |