missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GGH Protein

A cDNA sequence encoding the GGH was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11960-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918938

  • 103.00€ / 5 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

8836
Greater than 95% as determined by SDS-PAGE.
Research Use Only
GGH
Human
His
The GGH solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol.
GGH Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MRPHGDTAKK PIIGILMQKC RNKVMKNYGR YYIAASYVKY LESAGARVVP VRLDLTEKDY EILFKSINGI LFPGGSVDLR RSDYAKVAKI FYNLSIQSFD DGDYFPVWGT CLGFEELSLL ISGECLLTAT DTVDVAMPLN FTGGQLHSRM FQNFPTELLL SLAVEPLTAN FHKWSLSVKN FTMNEKLKKF FNVLTTNTDG KIEFISTMEG YKYPVYGVQW HPEKAPYEWK NLDGISHAPN AVKTAFYLAE FFVNEARKNN HHFKSESEEE KALIYQFSPI YTGNISSFQQ CYIFD
Videos