Learn More
enQuireBio™ Recombinant Human GFER Protein
A cDNA sequence encoding the GFER was constructed and used to recombinantly synthesize the protein.
Marca:  enQuireBio™ QP11954-2ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15908918
  • 137.00€ / 2 microgramos
Fecha estimada de envío: 29-05-2024
para ver el stock
Especificaciones
GFER Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAPGE RGRFHGGNLF FLPGGARSEM MDDLATDARG RGAGRRDAAA SASTPAQAPT SDSPVAEDAS RRRPCRACVD FKTWMRTQQK RDTKFREDCP PDREELGRHS WAVLHTLAAY YPDLPTPEQQ QDMAQFIHLF SKFYPCEECA EDLRKRLCRN HPDTRTRACF TQWLCHLHNE VNRKLGKPDF DCSKVDERWR DGWKDGSCD
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GFER protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT.