missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDNF Protein

A DNA sequence encoding the Homo sapiens (Human) GDNF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6078-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909819

  • 1926.00€ / 1 miligramo


Fecha estimada de envío
19-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000505.1
GDNF Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2668
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GDNF
Recombinant Protein
E. coli
SPDKQMAVLP RRERNRQAAA ANPENSRGKG RRGQRGKNRG CVLTAIHLNV TDLGLGYETK EELIFRYCSG SCDAAETTYD KILKNLSRNR RLVSDKVGQA CCRPIAFDDD LSFLDDNLVY HILRKHSAKR CGCI
Videos
Documentos