missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDI1 Protein
A cDNA sequence encoding the GDI1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11951-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GDI1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDEEYDV IVLGTGLTEC ILSGIMSVNG KKVLHMDRNP YYGGESSSIT PLEELYKRFQ LLEGPPESMG RGRDWNVDLI PKFLMANGQL VKMLLYTEVT RYLDFKVVEG SFVYKGGKIY KVPSTETEAL ASNLMGMFEK RRFRKFLVFV ANFDENDPKT FEGVDPQTTS MRDVYRKFDL GQDVIDFTGH ALALYRTDDY LDQPCLETVN RIKLYSESLA RYGKSPYLYP LYGLGELPQG FARLSAIYGG TYMLNKPVDD IIMENGKVVG VKSEGEVARC KQLICDPSYI PDRVRKAGQV IRIICILSHP IKNTNDANSC QIIIPQNQVN RKSDIYVCMI SYAHNVAAQG KYIAIASTTV ETTDPEKEVE PALELLEPID QKFVAISDLY EPIDDGCESQ VFCSCSYDAT THFETTCNDI KDIYKRMAGT AFDFENMKRK QNDVFGEAEQ | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GDI1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |