missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAS7 Protein
A cDNA sequence encoding the GAS7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11934-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GAS7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKPGMVPPPP GEESQTVILP PGWQSYLSPQ GRRYYVNTTT NETTWERPSS SPGIPASPGS HRSSLPPTVN GYHASGTPAH PPETAHMSVR KSTGDSQNLG SSSPSKKQSK ENTITINCVT FPHPDTMPEQ QLLKPTEWSY CDYFWADKKD PQGNGTVAGF ELLLQKQLKG KQMQKEMSEF IRERIKIEED YAKNLAKLSQ NSLASQEEGS LGEAWAQVKK SLADEAEVHL KFSAKLHSEV EKPLMNFREN FKKDMKKCDH HIADLRKQLA SRYASVEKAR KALTERQRDL EMKTQQLEIK LSNKTEEDIK KARRKSTQAG DDLMRCVDLY NQAQSKWFEE MVTTTLELER LEVERVEMIR QHLCQYTQLR HETDMFNQST VEPVDQLLRK VDPAKDRELW VREHKTGNIR PVDMEI | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GAS7 protein solution contains 20mM Tris-HCl, pH 8.0, 100mM NaCl, 2mM DTT and 10% glycerol. |