missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAP43 Protein
A cDNA sequence encoding the GAP43 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11931-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2596 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLCCMR RTKQVEKNDD DQKIEQDGIK PEDKAHKAAT KIQASFRGHI TRKKLKGEKK DDVQAAEAEA NKKDEAPVAD GVEKKGEGTT TAEAAPATGS KPDEPGKAGE TPSEEKKGEG DAATEQAAPQ APASSEEKAG SAETESATKA STDNSPSSKA EDAPAKEEPK QADVPAAVTA AAATTPAAED AAAKATAQPP TETGESSQAE ENIEAVDETK PKESARQDEG KEEEPEADQE HA | |
Greater than 90% as determined by SDS-PAGE. |
GAP43 Protein | |
Research Use Only | |
GAP43 | |
Human | |
His | |
The GAP43 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl and 10% glycerol. |