missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Gamma-secretase subunit APH-1A Protein

A DNA sequence encoding the Homo sapiens(Human) Gamma-secretase subunit APH-1A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10013-iv-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15932758

  • 845.00€ / 50 microgramos


Fecha estimada de envío
26-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001071096.1
Gamma-secretase subunit APH-1A Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
51107
Greater than 85% as determined by SDS-PAGE.
Research Use Only
APH1a
Recombinant Protein
Invitro-E. coli
MGAAVFFGCT FVAFGPAFAL FLITVAGDPL RVIILVAGAF FWLVSLLLAS VVWFILVHVT DRSDARLQYG LLIFGAAVSV LLQEVFRFAY YKLLKKADEG LASLSEDGRS PISIRQMAYV SGLSFGIISG VFSVINILAD ALGPGVVGIH GDSPYYFLTS AFLTAAIILL HTFWGVVFFD ACERRRYWAL GLVVGSHLLT SGLTFLNPWY EASLLPIYAV TVSMGLWAFI TAGGSLRSIQ RSLLCRR
Videos
Documentos