missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Gamma-secretase subunit APH-1A Protein
A DNA sequence encoding the Homo sapiens(Human) Gamma-secretase subunit APH-1A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10013-iv-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001071096.1 | |
Gamma-secretase subunit APH-1A Protein | |
Research Use Only | |
APH1a | |
Recombinant Protein | |
E. coli | |
MGAAVFFGCT FVAFGPAFAL FLITVAGDPL RVIILVAGAF FWLVSLLLAS VVWFILVHVT DRSDARLQYG LLIFGAAVSV LLQEVFRFAY YKLLKKADEG LASLSEDGRS PISIRQMAYV SGLSFGIISG VFSVINILAD ALGPGVVGIH GDSPYYFLTS AFLTAAIILL HTFWGVVFFD ACERRRYWAL GLVVGSHLLT SGLTFLNPWY EASLLPIYAV TVSMGLWAFI TAGGSLRSIQ RSLLCRR | |
Greater than 85% as determined by SDS-PAGE. |
51107 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |