missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Gamma-crystallin D Protein
A DNA sequence encoding the Homo sapiens (Human) Gamma-crystallin D, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5871-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_008822.2 | |
Gamma-crystallin D Protein | |
Research Use Only | |
CRYGD | |
Recombinant Protein | |
E. coli | |
GKITLYEDRG FQGRHYECSS DHPNLQPYLS RCNSARVDSG CWMLYEQPNY SGLQYFLRRG DYADHQQWMG LSDSVRSCRL IPHSGSHRIR LYEREDYRGQ MIEFTEDCSC LQDRFRFNEI HSLNVLEGSW VLYELSNYRG RQYLLMPGDY RRYQDWGATN ARVGSLRRVI DFS | |
Greater than 80% as determined by SDS-PAGE. |
1421 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |