missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galectin-1 / LGALS1 Protein

A DNA sequence encoding the Human Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10191-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923048

  • 114.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 11-04-2019
Añadir a la cesta

Especificación

Especificación

NP_002296.1
Galectin-1 / LGALS1 Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD
3956
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
LGALS1
The biological activity determined by a chemotaxis bioassay using human blood monocytes is in a concentration range of 1.0-10 ng/ml.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
Videos