missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FUT3 Protein

A cDNA sequence encoding the FUT3 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11911-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918778

  • 103.00€ / 2 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

FUT3 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The FUT3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSRVSRDDA TGSPRAPSGS SRQDTTPTRP TLLILLWTWP FHIPVALSRC SEMVPGTADC HITADRKVYP QADTVIVHHW DIMSNPKSRL PPSPRPQGQR WIWFNLEPPP NCQHLEALDR YFNLTMSYRS DSDIFTPYGW LEPWSGQPAH PPLNLSAKTE LVAWAVSNWK PDSARVRYYQ SLQAHLKVDV YGRSHKPLPK GTMMETLSRY KFYLAFENSL HPDYITEKLW RNALEAWAVP VVLGPSRSNY ERFLPPDAFI HVDDFQSPKD LARYLQELDK DHARYLSYFR WRETLRPRSF SWALDFCKAC WKLQQESRYQ TVRSIAAWFT
Videos